Tisk…
Cleanroom gloves, neoprene, sterile, BioClean Suprene™
Dodavatel: Nitritex
BSNS-65 BSNS-70 BSNS-75 BSNS-60 BSNS-80 BSNS-90 BSNS-85
112-3338EA 19660 CZK
112-3338 112-3339 112-3340 112-3337 112-3341 112-3343 112-3342
Cleanroom gloves, neoprene, sterile, BioClean Suprene™
Rukavice
These anatomically shaped gloves are processed to ensure ISO class 4 and EU GMP grade A compatibility. Thanks to their excellent strength and stretch these gloves are the ideal choice for use within isolators.
  • Sterile, powder-free
  • Latex-free to eliminate type 1 latex allergies
  • Beaded cuff and textured surface for increased tactility
  • Double-donnable with good grip
  • Neoprene gives strength and durability
  • Excellent ESD properties
Suitable for use in class 10, ISO 4 or M2.5 and EU GMP grade A cleanrooms.
EN 374, EN 455, ASTM D 6978-05 tested; Cat. III, AQL 1,5
Colour: Light green
EN 374-1/2/3, EN 455 (Parts 1, 2 & 3), ASTM D 6978-05 tested; Cat. III, AQL 1,5
Certifikace: EN 455-2, EN ISO 374-1 type C, EN ISO 374-5 VIRUS, EN 421; Cat. III
Food contact approved
Tested against permeation standard ASTM D6978-05 for cytotoxic drugs.
Balení: One cuffed pair is sealed in an inner plastic wallet which is then sealed in an EasyTear™ PE pouch. Ten pouches are sealed in an outer PE bag and the carton liner contains 20 outer bags.