Tisk…
Blades for cutters
Dodavatel: THEODOR KIHLER BURO EINRICHTUN
CB9 CB18
233-2311EA 0 CZK
233-2311 233-2321
Blades for cutters
Žiletky a čepele
Spare blades for Edding cutters.
  • High-quality
  • One blade corresponds to multiple cutters
Remove the tail piece, lift the flap and pull out the blade holder. Carefully remove the remaining blade, insert the new blade and slide it back into the cutter. After that, put the tail piece back on.