Tisk…
Multi purpose spray, WD-40
Dodavatel: KOEHLER TECHNISCHE PRODUKTEN
K-1595 K-1596
291-1253EA 515 CZK
291-1253 KOEHK-1596
Multi purpose spray, WD-40
Lubrikanty
This multifunctional product creates a perfect barrier against moisture by infiltrating it. This means that electrical and electronic mechanisms always remain protected and leakage currents are avoided. One of the main components is an active lubricant, which makes moving parts easier to move while eliminating squeaks and creaks.
  • Silicone-free, resin-free and PTFE-free
  • Not conductive
  • Compatible with various metals
  • 360° valve for use from any position
  • 2-way spray system: Can be used over a wide area or precisely at one point
Dissolves stubborn dirt, oil, grease and glue residues and makes them easy to remove. Leaves a nourishing protective layer. Also loosens stuck mechanisms and jammed metal parts by infiltrating layers of rust to loosen them. Thanks to its corrosion-resistant formula, it offers long-lasting protection against rust and all forms of oxidation on metal surfaces.