Tisk…
Occupational shoes, slip-on, HACCP, Air Cushion, 2370, Abeba
Dodavatel: Abeba
2370/35 2370/36 2370/37 2370/38 2370/39 2370/40 2370/41 2370/42 2370/43 2370/44 2370/45 2370/46 2370/47
114-0522EA 2160 CZK
114-0522 114-0523 114-0524 114-0525 114-0526 114-0527 114-0528 114-0529 114-0530 114-0531 114-0532 114-0533 114-0534
Occupational shoes, slip-on, HACCP, Air Cushion, 2370, Abeba
Obuv
Anti-static occupational shoes made from microfibre upper and PU sole resistant to hydrocarbons.
  • Air-cushioned PU sole with high slip resistance
  • Integrated insole
  • Sanitized® treatment reduces odour and bacterial growth
  • Washable at 60 °C
Suitable for use in kitchens.
Colour: White.
Certifikace: HACCP; EN 20347:2012; O1, SRA.