Tisk…
Chemical cabinets for small containers
Dodavatel: LACONT UMWELTTECHNIK
B80-6207-A B80-6208-A
LACOB80-6207-AEA 64810 CZK
LACOB80-6207-A LACOB80-6208-A
Chemical cabinets for small containers
Nábytek kancelářský Skříně pro zásoby
Chemical and poison cabinets are ideal for safe and regular storage of non-flammable hazardous substances, chemicals and flammable liquids in workspaces.
  • Doors with central cylinder lock and ventilation slots
  • Central DN 80 ventilation connection for exhaust air extraction of both cabinets on roof
  • Available with six door colours
  • Automatic thermal closing mechanism in case of fire
Cabinets are made of single-wall body made from high-quality powder-coated sheet steel in RAL 7035 - light grey Model GL with acrylic glass doors. It also contains small safety cabinet with separate cylinder lock.
Tray-shaped shelves are non-tilt and can hold maximum load of 50 kg/shelf. It is height-adjustable at intervals of 25 mm. Ground connection for avoidance of sources of ignition.
Chemical cabinet has 3× tray-shaped shelves whereas small safety cabinet has base sump tray with sump capacity of 11 litres.
Certifikace: Conforms with DIN EN 14470-1, TRBS 2153, TRGS 514, TRGS 526, BGR 120, TRbF 20-L.