Tisk…

Phenyl bromoacetate ≥98%

Dodavatel: Thermo Scientific

Synonyms: Bromoacetic acid Phenyl ester, Phenyl 2-bromoacetate

H60464.06 H60464.18
H60464.06EA 1280 CZK
H60464.06 H60464.18
Phenyl bromoacetate ≥98%
Phenyl bromoacetate
Vzorec: C₈H₇BrO₂
MW: 215,04 g/mol
Bod varu: 134 °C
Bod tání: 31…33 °C
Hustota: 1,508
Bod vzplanutí: 113 °C (235 °F)
Storage Temperature: Ambient
MDL Number: MFCD00192391
CAS číslo: 620-72-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR