Tisk…

(±)-2-Amino-2-(4-chlorophenyl)acetic acid ≥98%

Dodavatel: Thermo Fisher Scientific
Varování

Synonyms: DL-2-(4-Chlorophenyl)glycine, H-DL-Phg(4-Cl)-OH, DL-alpha-Amino-4-chlorophenylacetic acid, 4-Chloro-DL-2-phenylglycine, 4-Chloro-DL-phenylglycine, 2-Amino-2-(4-chlorophenyl)acetic acid

H59921.14 H59921.22
H59921.14EA 3570 CZK
H59921.14 H59921.22
(±)-2-Amino-2-(4-chlorophenyl)acetic acid ≥98%
(±)-2-Amino-2-(4-chlorophenyl)acetic acid
Vzorec: C₈H₈ClNO₂
Storage Temperature: Ambient
MDL Number: MFCD00049328
CAS číslo: 6212-33-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR