Tisk…

Ethyl-2-aminothiophene-3-carboxylate 98%

Dodavatel: Thermo Scientific
Varování

Synonyms: Ethyl 2-aminothiophene-3-carboxylate

H61559.14 H61559.22
H61559.14EA 4430 CZK
H61559.14 H61559.22
Ethyl-2-aminothiophene-3-carboxylate 98%
Ethyl-2-aminothiophene-3-carboxylate
Vzorec: C₇H₉NO₂S
MW: 171,22 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01566303
CAS číslo: 31891-06-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR