Malonyl dichloride ≥97%
Dodavatel: Thermo Scientific
|
Nebezpečí
|
H59868.14
H59868.22
H59868.14EA
4810
CZK
H59868.14
H59868.22
Malonyl dichloride ≥97%
Malonyl dichloride
Purity: 97%
CAS No.: 1663-67-8
Molecular Formula: C3H2Cl2O2
Formula Weight: 140.95
Flash Point: 47°(117°F)
Refractive Index: 1.4650
Boiling Point: 53-55°/19mm
Beilstein Reference: 774044
UN No.: 2920
MDL No.: MFCD00000735
Vzorec:
C₃H₂Cl₂O₂ MW: 140,95 g/mol Bod varu: 53…55 °C (19 mmHg) Hustota: 1,449 Bod vzplanutí: 47 °C (117 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000735 CAS číslo: 1663-67-8 EINECS: 216-772-9 UN: 2920 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...