Benzyldimethyldodecylammonium chloride ≥98%
Dodavatel: Thermo Scientific
|
Nebezpečí
|
Synonyms:
Benzyldimethyldodecylammonium chloride, N-Benzyl-N,N-dimethyldodecan-1-aminium chloride, Dodecyldimethylbenzylammonium chloride, Benzyldodecyldimethylammonium chloride dihydrate
H60196.06
H60196.18
H60196.06EA
1880
CZK
H60196.06
H60196.18
Benzyldimethyldodecylammonium chloride ≥98%
Benzyldimethyldodecylammonium chloride
Vzorec:
C₂₁H₃₈ClN MW: 339,99 g/mol Bod tání: 60 °C Storage Temperature: Ambient |
MDL Number:
MFCD00137276 CAS číslo: 139-07-1 EINECS: 205-351-5 UN: 3261 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...