4-Bromo-2-methylbenzaldehyde 95%
Dodavatel: Thermo Scientific
|
Varování
|
H61314.03
H61314.06
H61314.03EA
5800
CZK
H61314.03
H61314.06
4-Bromo-2-methylbenzaldehyde 95%
4-Bromo-2-methylbenzaldehyde
Vzorec:
C₈H₇BrO Storage Temperature: Ambient |
MDL Number:
MFCD07787171 CAS číslo: 24078-12-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...