Tisk…
1-Ethyl-3-methylimidazolium acetate 97%
Dodavatel: Thermo Fisher Scientific
Synonyms: 1-Ethyl-3-methylimidazolium Acetate
H59277.06 H59277.18
H59277.06EA 4050 CZK
H59277.06 H59277.18
1-Ethyl-3-methylimidazolium acetate 97%
1-Ethyl-3-methylimidazolium acetate
Vzorec: C₈H₁₄N₂O₂
MW: 170.21
MDL Number: MFCD06798186
CAS číslo: 143314-17-4