Tisk…

Polymyxin B sulphate

Dodavatel: Cayman Chemical

14157-500 14157-1 14157-5
CAYM14157-500EA 1540 CZK
CAYM14157-500 CAYM14157-1 CAYM14157-5
Polymyxin B sulphate
Polymyxin B sulphate
Vzorec: C₅₅H₉₆N₁₆O₁₃·2H₂SO₄
MW: 1385.61 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00131991
CAS číslo: 1405-20-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR