Tisk…

4-Bromo-2,6-difluorobenzyl alcohol ≥97%

Dodavatel: Thermo Fisher Scientific
Varování

Synonyms: (4-Bromo-2,6-difluorophenyl)methanol

H61623.03 H61623.06
H61623.03EA 1330 CZK
H61623.03 H61623.06
4-Bromo-2,6-difluorobenzyl alcohol ≥97%
4-Bromo-2,6-difluorobenzyl alcohol
Vzorec: C₇H₅BrF₂O
MW: 223.02
Bod tání: 76…81 °C
Storage Temperature: Ambient
MDL Number: MFCD03094461
CAS číslo: 162744-59-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR