Tisk…

Butyl levulinate ≥98%

Dodavatel: Thermo Fisher Scientific

Synonyms: Levulinic acid butyl ester, 4-Oxovaleric acid butyl ester, Butyl 4-oxovalerate

H59274.AE H59274.AP
H59274.AEEA 745 CZK
H59274.AE H59274.AP
Butyl levulinate ≥98%
Butyl levulinate
Vzorec: C₉H₁₆O₃
Bod varu: 106…108 °C (5,5 mmHg)
Hustota: 0,974
Bod vzplanutí: 92 °C (198 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009449
CAS číslo: 2052-15-5
EINECS: 218-143-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR