Gemfibrozil
Dodavatel: Cayman Chemical
14835-25
14835-1
14835-5
14835-10
CAYM14835-25EA
7730
CZK
CAYM14835-25
CAYM14835-1
CAYM14835-5
CAYM14835-10
Gemfibrozil
Gemfibrozil
Vzorec:
C₁₅H₂₂O₃ Bod varu: 395 °C (1013 hPa) Bod tání: 59…61 °C |
MDL Number:
MFCD00079335 CAS číslo: 25812-30-0 EINECS: 247-280-2 Merck Index: 12,04394 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...