Tisk…

4-Fluorobenzoylacetonitrile ≥97%

Dodavatel: Thermo Scientific
Varování

Synonyms: 3-(4-Fluorophenyl)-3-oxopropanenitrile

H61988.03
H61988.03EA 1210 CZK
H61988.03
4-Fluorobenzoylacetonitrile ≥97%
4-Fluorobenzoylacetonitrile

Purity: 97% CAS No.: 4640-67-9 Molecular Formula: C9H6FNO2 Formula Weight: 163.15 Physical Form: Solid Appearance: Off-white to pale yellow Melting Point: 84-88° UN No.: 3439 MDL No.: MFCD00662062

Vzorec: C₉H₆FNO
MW: 163,15 g/mol
Bod tání: 84…88 °C
Storage Temperature: Ambient
MDL Number: MFCD00662062
CAS číslo: 4640-67-9
UN: 3439
ADR: 6.1,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR