Tisk…

Methyl (±)-lactate ≥99%

Dodavatel: Thermo Fisher Scientific
Varování

Synonyms: Methyl Lactate, Methyl DL-lactate, Lactic acid methyll ester, Methyl (±)-2-hydroxypropionate

H60455.AA H60455.AC
H60455.AAEA 2260 CZK
H60455.AA H60455.AC
Methyl (±)-lactate ≥99%
Methyl (±)-lactate
Vzorec: C₄H₈O₃
Bod varu: 35 °C (1013 hPa)
Hustota: 1,093
Bod vzplanutí: 51 °C (124 °F)
Storage Temperature: Ambient
MDL Number: MFCD00066367
CAS číslo: 547-64-8
EINECS: 208-930-0
UN: 3272
ADR: 3,III
Merck Index: 12,06167

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR