4-Nitro-α,α,α-trifluoro-o-toluic acid ≥97%
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
4-Nitro-2-(trifluoromethyl)benzoic acid
H61452.03
H61452.06
H61452.03EA
1890
CZK
H61452.03
H61452.06
4-Nitro-α,α,α-trifluoro-o-toluic acid ≥97%
4-Nitro-α,α,α-trifluoro-o-toluic acid
Vzorec:
C₈H₄F₃NO₄ MW: 235,12 g/mol Bod tání: 128…132 °C Storage Temperature: Ambient |
MDL Number:
MFCD04039246 CAS číslo: 320-37-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...