Dimethyl fumarate
Dodavatel: Cayman Chemical
Synonyms:
Fumaric acid dimethyl ester
14714-50
14714-100
CAYM14714-50EA
1750
CZK
CAYM14714-50
CAYM14714-100
Dimethyl fumarate
Dimethyl fumarate
Vzorec:
C₆H₈O₄ MW: 144,13 g/mol Bod varu: 192 °C (1013 hPa) Bod tání: 102 °C Hustota: 1,37 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00064438 CAS číslo: 624-49-7 EINECS: 210-849-0 Merck Index: 13,04308 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...