1-Bromo-3-chloro-2-fluorobenzene 95%
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
3-Chloro-2-fluorobromobenzene
H61622.14
H61622.22
H61622.14EA
1710
CZK
H61622.14
H61622.22
1-Bromo-3-chloro-2-fluorobenzene 95%
1-Bromo-3-chloro-2-fluorobenzene
Vzorec:
C₆H₃BrClF MW: 209,44 g/mol Bod tání: 42…46 °C Storage Temperature: Ambient |
MDL Number:
MFCD03412182 CAS číslo: 144584-65-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...