5-Fluoro-2-nitroaniline 97%
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
H61806.06
H61806.14
H61806.22
H61806.06EA
1240
CZK
H61806.06
H61806.14
H61806.22
5-Fluoro-2-nitroaniline 97%
5-Fluoro-2-nitroaniline
Vzorec:
C₆H₅FN₂O₂ MW: 156.12 Storage Temperature: Ambient |
MDL Number:
MFCD00034065 CAS číslo: 2369-11-1 EINECS: 219-128-5 UN: 2811 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...