2-Bromo-p-toluic acid ≥97%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
H60413.03
H60413.06
H60413.03EA
1690
CZK
H60413.03
H60413.06
2-Bromo-p-toluic acid ≥97%
2-Bromo-p-toluic acid
Vzorec:
C₈H₇BrO₂ MW: 215.05 Bod tání: 143…147 °C Storage Temperature: Ambient |
MDL Number:
MFCD00045797 CAS číslo: 7697-27-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...