Tisk…

Ethyl-2-piperidinecarboxylate, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: Ethyl 2-Piperidinecarboxylate, Ethyl pipecolinate

198803-25G 198803-5G
SIAL198803-25GEA 7420 CZK
SIAL198803-25G SIAL198803-5G
Ethyl-2-piperidinecarboxylate, Sigma-Aldrich®
Ethyl-2-piperidinecarboxylate
Vzorec: C₈H₁₅NO₂
MW: 157,21 g/mol
MDL Number: MFCD00005980
CAS číslo: 15862-72-3
EINECS: 239-990-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR