Tisk…

Ethyl-2-piperidinecarboxylate ≥98%

Dodavatel: Thermo Scientific
Varování

Synonyms: Ethyl 2-Piperidinecarboxylate, Ethyl pipecolinate

L10532.03 L10532.06 L10532.14
L10532.03EA 721 CZK
L10532.03 L10532.06 L10532.14
Ethyl-2-piperidinecarboxylate ≥98%
Ethyl-2-piperidinecarboxylate

Purity: 98+% CAS No.: 15862-72-3 Molecular Formula: C8H15NO2 Formula Weight: 157.21 Flash Point: 46°(114°F) Refractive Index: 1.4560 Boiling Point: 216-217° Beilstein Reference: 81694 UN No.: 1993 MDL No.: MFCD00005980

Vzorec: C₈H₁₅NO₂
MW: 157,21 g/mol
Bod varu: 216…217 °C
Hustota: 1,006
Bod vzplanutí: 46 °C (114 °F)
Storage Temperature: Ambient
MDL Number: MFCD00005980
CAS číslo: 15862-72-3
EINECS: 239-990-6
ADR: 3,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR