2,5-Di-tert-butylaniline, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ100
275735-10G
275735-50G
SIAL275735-10GEA
6070
CZK
SIAL275735-10G
SIAL275735-50G
2,5-Di-tert-butylaniline, Sigma-Aldrich®
2,5-Di-tert-butylaniline
Vzorec:
C₁₂H₁₉N |
MDL Number:
MFCD00010338 CAS číslo: 21860-03-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...