Tisk…

4,4'-Diisopropylbiphenyl ≥97%

Dodavatel: Thermo Fisher Scientific

B20779.03 B20779.06
B20779.03EA 1080 CZK
B20779.03 B20779.06
4,4'-Diisopropylbiphenyl ≥97%
4,4'-Diisopropylbiphenyl

Purity: 97% CAS No.: 18970-30-4 Molecular Formula: C18H22 Formula Weight: 238.37 Melting Point: 66° MDL No.: MFCD00043533

Vzorec: C₁₈H₂₂
Bod tání: 66 °C
Storage Temperature: Ambient
MDL Number: MFCD00043533
CAS číslo: 18970-30-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR