4-Bromo-1-chloro-2-ethylbenzene ≥98%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
B25613.06
B25613.14
B25613.06EA
1900
CZK
B25613.06
B25613.14
4-Bromo-1-chloro-2-ethylbenzene ≥98%
4-Bromo-1-chloro-2-ethylbenzene
Purity: 98+%
CAS No.: 289039-22-1
Molecular Formula: C8H8BrCl
Formula Weight: 219.51
MDL No.: MFCD00672954
Vzorec:
C₈H₈BrCl Storage Temperature: Ambient |
MDL Number:
MFCD00672954 CAS číslo: 289039-22-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...