Tisk…

4-(Trimethylsilyl)-3-butyn-2-ol, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: 4-(Trimethylsilyl)but-3-yn-2-ol

M-Clarity™ quality level = MQ100

666955-25G 666955-5G
SIAL666955-25GEA 9580 CZK
SIAL666955-25G SIAL666955-5G
4-(Trimethylsilyl)-3-butyn-2-ol, Sigma-Aldrich®
4-(Trimethylsilyl)-3-butyn-2-ol
Vzorec: C₇H₁₄OSi
MW: 142,27 g/mol
MDL Number: MFCD00190213
CAS číslo: 6999-19-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR