4-Phenyl-1-butyne ≥98%
Dodavatel: Thermo Scientific
L12946.06
L12946.14
L12946.06EA
3300
CZK
L12946.06
L12946.14
4-Phenyl-1-butyne ≥98%
4-Phenyl-1-butyne
Purity: 98%
CAS No.: 16520-62-0
Molecular Formula: C10H10
Formula Weight: 130.19
Refractive Index: 1.5210
Boiling Point: 189-190°
MDL No.: MFCD00041667
Vzorec:
C₁₀H₁₀ MW: 130,19 g/mol Bod varu: 189…190 °C Hustota: 0,926 Storage Temperature: Ambient |
MDL Number:
MFCD00041667 CAS číslo: 16520-62-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...