Tisk…

Ammonium perfluorooctanoate, Sigma-Aldrich®

Dodavatel: Merck

77262-10G 77262-50G
SIAL77262-10GEA 3010 CZK
SIAL77262-10G SIAL77262-50G
Ammonium perfluorooctanoate, Sigma-Aldrich®
Ammonium perfluorooctanoate
Vzorec: C₈H₄F₁₅NO₂
MDL Number: MFCD00042599
CAS číslo: 3825-26-1
EINECS: 223-320-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR