Perfluorodecanoic acid ≥97%
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
B22292.06
B22292.14
B22292.06EA
2130
CZK
B22292.06
B22292.14
Perfluorodecanoic acid ≥97%
Perfluorodecanoic acid
Purity: 97%
CAS No.: 335-76-2
Molecular Formula: C10HF19O2
Formula Weight: 514.09
Melting Point: 77-82°
Boiling Point: 218°
UN No.: 2923
MDL No.: MFCD00004175
Vzorec:
C₁₀HF₁₉O₂ Bod varu: 218 °C Bod tání: 77…82 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004175 CAS číslo: 335-76-2 EINECS: 206-400-3 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...