5-Cyano-2-methylpyridine ≥99%
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
6-Methylnicotinonitrile, 5-Cyano-2-picoline
L09875.03
L09875.06
L09875.03EA
1020
CZK
L09875.03
L09875.06
5-Cyano-2-methylpyridine ≥99%
5-Cyano-2-methylpyridine
Purity: 99%
CAS No.: 3222-48-8
Molecular Formula: C7H6N2
Formula Weight: 118.14
Melting Point: 83-87°
Boiling Point: 216-217°
MDL No.: MFCD00038042
Vzorec:
C₇H₆N₂ MW: 118,14 g/mol Bod varu: 216…217 °C Bod tání: 83…87 °C Storage Temperature: Ambient |
MDL Number:
MFCD00038042 CAS číslo: 3222-48-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...