Sodium stannate hydrate, Reagent Grade
Dodavatel: Thermo Fisher Scientific
|
Varování
|
Synonyms:
Sodium tin(IV) oxide hydrate
14021.22
14021.36
14021.22EA
1420
CZK
14021.22
14021.36
Sodium stannate hydrate, Reagent Grade
Sodium stannate hydrate
Sodium tin(IV) oxide is used as a mordant, fire retardant textiles.
CAS No.: 12058-66-1
Molecular Formula: Na2SnO3.xH2O
Formula Weight: 212.67(anhy)
Physical Form: Crystalline
Sensitivity: Air Sensitive
Odour: Odourless
Merck Reference: 14,8677
MDL No.: MFCD00014241
Vzorec:
Na₂O₃Sn·H₂O MW: 212.67 g/mol (anhy) Bod tání: 140 °C (rozkl.) Storage Temperature: Ambient |
MDL Number:
MFCD00014241 CAS číslo: 12058-66-1 EINECS: 235-030-5 Merck Index: 14,8677 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...