Tisk…

3-(4-Chlorophenoxy)propionic acid, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: 3-(4-Chlorophenoxy)propanoic acid

M-Clarity™ quality level = MQ100

656305-25G 656305-5G
SIAL656305-25GEA 8530 CZK
SIAL656305-25G SIAL656305-5G
3-(4-Chlorophenoxy)propionic acid, Sigma-Aldrich®
3-(4-Chlorophenoxy)propionic acid
Vzorec: C₉H₉ClO₃
MW: 200,6190 g/mol
MDL Number: MFCD00043504
CAS číslo: 3284-79-5
EINECS: 221-932-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR