Tisk…

(1-Propynyl)tributyltin, Sigma-Aldrich®

Dodavatel: Merck
Nebezpečí


M-Clarity™ quality level = MQ100

499862-1G 499862-5G
SIAL499862-1GEA 3320 CZK
SIAL499862-1G SIAL499862-5G
(1-Propynyl)tributyltin, Sigma-Aldrich®
(1-Propynyl)tributyltin
Vzorec: C₁₅H₃₀Sn
MW: 329,12 g/mol
MDL Number: MFCD01863650
CAS číslo: 64099-82-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR