(1-Propynyl)tributyltin 96%
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
L20201.03
L20201.06
L20201.03EA
2000
CZK
L20201.03
L20201.06
(1-Propynyl)tributyltin 96%
(1-Propynyl)tributyltin
Purity: 96%
CAS No.: 64099-82-7
Molecular Formula: C15H30Sn
Formula Weight: 329.12
Storage Temperature: 2°C to 8°C
Flash Point:
Vzorec:
C₁₅H₃₀Sn Bod varu: 277 °C Hustota: 1,082 Bod vzplanutí: >110 °C (230 °F) |
MDL Number:
MFCD01863650 CAS číslo: 64099-82-7 ADR: 6.1,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...