Tisk…

D-Phenylalaninol ≥98%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

L09697.03 L09697.MD
L09697.03EA 1260 CZK
L09697.03 L09697.MD
D-Phenylalaninol ≥98%
D-Phenylalaninol

Purity: 98% CAS No.: 5267-64-1 Molecular Formula: C9H13NO Formula Weight: 151.21 Melting Point: 89-94° Beilstein Reference: 4665408 UN No.: 3259 MDL No.: MFCD00064298

Vzorec: C₉H₁₃NO
Bod tání: 89…94 °C
Storage Temperature: Ambient
MDL Number: MFCD00064298
CAS číslo: 5267-64-1
EINECS: 226-086-1
UN: 3259
ADR: 8,III

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR