tert-Butylacetyl chloride, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
3,3-Dimethylbutyryl chloride
M-Clarity™ quality level = MQ200
B88802-100G
B88802-25G
SIALB88802-100GEA
5690
CZK
SIALB88802-100G
SIALB88802-25G
tert-Butylacetyl chloride, Sigma-Aldrich®
tert-Butylacetyl chloride
Vzorec:
C₆H₁₁ClO MW: 134,61 g/mol |
MDL Number:
MFCD00000737 CAS číslo: 7065-46-5 EINECS: 230-356-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...