2-Ethylbutyryl chloride ≥99%
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
B23774.14
B23774.22
B23774.14EA
2830
CZK
B23774.14
B23774.22
2-Ethylbutyryl chloride ≥99%
2-Ethylbutyryl chloride
Purity: 99%
CAS No.: 2736-40-5
Molecular Formula: (CH3CH2)2CHCOCl
Formula Weight: 134.61
Flash Point: 33°(91°F)
Refractive Index: 1.4250
Odour: Acrid
Boiling Point: 137-139°
UN No.: 3094
MDL No.: MFCD00018812
Vzorec:
C₆H₁₁ClO Bod varu: 137…139 °C Hustota: 0,980 Bod vzplanutí: 33 °C (91 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00018812 CAS číslo: 2736-40-5 EINECS: 220-359-9 UN: 2920 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...