3-Methyl-1,5-pentanediol ≥99%
Dodavatel: Thermo Fisher Scientific
Synonyms:
3-Methylpentane-1,5-diol
B20546.06
B20546.14
B20546.06EA
2230
CZK
B20546.06
B20546.14
3-Methyl-1,5-pentanediol ≥99%
3-Methyl-1,5-pentanediol
Purity: 99%
CAS No.: 4457-71-0
Molecular Formula: HOCH2CH2CH(CH3)CH2CH2OH
Formula Weight: 118.18
Flash Point: 143°(289°F)
Refractive Index: 1.454
Beilstein Reference: 1697331
MDL No.: MFCD00039547
Vzorec:
C₆H₁₄O₂ MW: 118.18 g/mol Hustota: 0.97 Bod vzplanutí: 143 °C (289 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00039547 CAS číslo: 4457-71-0 EINECS: 224-709-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...