Tisk…

2-Amino-4,6-dimethylpyridine, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

A51807-25G A51807-5G
SIALA51807-25GEA 9410 CZK
SIALA51807-25G SIALA51807-5G
2-Amino-4,6-dimethylpyridine, Sigma-Aldrich®
2-Amino-4,6-dimethylpyridine
Vzorec: C₇H₁₀N₂
MW: 122,1700 g/mol
MDL Number: MFCD00006322
CAS číslo: 5407-87-4
EINECS: 226-470-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR