Tisk…

4-Bromo-1-butyne ≥97%

Dodavatel: Thermo Scientific
Nebezpečí

H53367.06 H53367.14
H53367.06EA 3990 CZK
H53367.06 H53367.14
4-Bromo-1-butyne ≥97%
4-Bromo-1-butyne

Purity: 97% CAS No.: 38771-21-0 Molecular Formula: C4H5Br Formula Weight: 132.99 Storage Temperature: 2°C to 8°C Flash Point: 24°(75°F) Refractive Index: 1.481 Boiling Point: 110° UN No.: 1992 MDL No.: MFCD10000883

Vzorec: C₄H₅Br
MW: 132,99 g/mol
Bod varu: 110 °C
Hustota: 1,417
Bod vzplanutí: 24 °C (75 °F)
MDL Number: MFCD10000883
CAS číslo: 38771-21-0
ADR: 3,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR