Tisk…

1,4-Bis(2-hydroxyisopropyl)benzene, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: 1,4-Bis(alpha-hydroxyisopropyl)benzene, alpha,alpha'-Dihydroxy-1,4-diisopropylbenzene, 2,2'-(p-Phenylene)di-2-propanol, 2,2'-(1,4-Phenylene)bis(propan-2-ol), alpha,alpha,alpha',alpha'-Tetramethyl-1,4-benzenedimethanol

M-Clarity™ quality level = MQ100

255149-25G 255149-5G
SIAL255149-25GEA 20250 CZK
SIAL255149-25G SIAL255149-5G
1,4-Bis(2-hydroxyisopropyl)benzene, Sigma-Aldrich®
1,4-Bis(2-hydroxyisopropyl)benzene
Vzorec: C₁₂H₁₈O₂
MW: 194,28 g/mol
MDL Number: MFCD00009827
CAS číslo: 2948-46-1
EINECS: 220-964-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR