1,4-Bis(2-hydroxyisopropyl)benzene ≥97%
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
1,4-Bis(alpha-hydroxyisopropyl)benzene, alpha,alpha'-Dihydroxy-1,4-diisopropylbenzene, 2,2'-(p-Phenylene)di-2-propanol, 2,2'-(1,4-Phenylene)bis(propan-2-ol), alpha,alpha,alpha',alpha'-Tetramethyl-1,4-benzenedimethanol
B21472.03
B21472.06
B21472.03EA
1150
CZK
B21472.03
B21472.06
1,4-Bis(2-hydroxyisopropyl)benzene ≥97%
1,4-Bis(2-hydroxyisopropyl)benzene
Purity: 97%
CAS No.: 2948-46-1
Molecular Formula: C12H18O2
Formula Weight: 194.28
Melting Point: 142-146°
Boiling Point: 167-168°/12mm
MDL No.: MFCD00009827
Vzorec:
C₁₂H₁₈O₂ MW: 194,28 g/mol Bod varu: 167…168 °C (12 mmHg) Bod tání: 142…146 °C Storage Temperature: Ambient |
MDL Number:
MFCD00009827 CAS číslo: 2948-46-1 EINECS: 220-964-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...