Tisk…

16-Mercaptopalmitic acid, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ200

448303-1G 448303-5G
SIAL448303-1GEA 6610 CZK
SIAL448303-1G SIAL448303-5G
16-Mercaptopalmitic acid, Sigma-Aldrich®
16-Mercaptopalmitic acid
Vzorec: C₁₆H₃₂O₂S
MDL Number: MFCD00674018
CAS číslo: 69839-68-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR