Tisk…

1-Isopropyl-4-piperidone ≥99%

Dodavatel: Thermo Fisher Scientific
Varování

A11480.09 A11480.18 A11480.30
A11480.09EA 841 CZK
A11480.09 A11480.18 A11480.30
1-Isopropyl-4-piperidone ≥99%
1-Isopropyl-4-piperidone

Purity: 99% CAS No.: 5355-68-0 Molecular Formula: C8H15NO Formula Weight: 141.22 Refractive Index: 1.4655 Boiling Point: 100-101°/27mm MDL No.: MFCD00038035

Vzorec: C₈H₁₅NO
Bod varu: 100…101 °C (27 mmHg)
Hustota: 0,950
Storage Temperature: Ambient
MDL Number: MFCD00038035
CAS číslo: 5355-68-0
EINECS: 226-339-6

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR