Chlorodimethyloctadecylsilane ≥95%
Dodavatel: Thermo Scientific
|
Nebezpečí
|
42414.06
42414.14
42414.22
42414.36
42414.06EA
1210
CZK
42414.06
42414.14
42414.22
42414.36
Chlorodimethyloctadecylsilane ≥95%
Chlorodimethyloctadecylsilane
Purity: 95%
CAS No.: 18643-08-8
Molecular Formula: C20H43ClSi
Formula Weight: 347.10
Flash Point: 200°(392°F)
Physical Form: Solid
Density: 0.850
Refractive Index: 1.4530
Sensitivity: Moisture Sensitive
Appearance: White
Odour: Acrid
Melting Point: 28-32°
Boiling Point: 184-186°/0.2mm
Beilstein Reference: 2246012
UN No.: 3261
MDL No.: MFCD00000504
Preparation of hydrophobic surfaces, chain terminator
Vzorec:
C₂₀H₄₃ClSi MW: 347,10 g/mol Bod varu: 184…186 °C (0,2 mmHg) Bod tání: 28…32 °C Hustota: 0,850 Bod vzplanutí: 200 °C (392 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000504 CAS číslo: 18643-08-8 EINECS: 242-472-2 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...