Tisk…

Zinc chloride hydrate ≥99.99% (metals basis)

Dodavatel: Thermo Scientific
Nebezpečí

41247.09 41247.18 41247.30
41247.09EA 1720 CZK
41247.09 41247.18 41247.30
Zinc chloride hydrate ≥99.99% (metals basis)
Zinc chloride hydrate

Purity: 99.99% CAS No.: 7646-85-7 Molecular Formula: Cl2Zn.xH2O Formula Weight: 136.29(anhy) Physical Form: Crystalline Aggregates Appearance: White Odour: Odourless Merck Reference: 14,10132 UN No.: 3260 MDL No.: MFCD00011295

Vzorec: Cl₂Zn·H₂O
Storage Temperature: Ambient
MDL Number: MFCD00011295
CAS číslo: 7646-85-7
EINECS: 231-592-0
UN: 3260
ADR: 8,III
Merck Index: 14,10132

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR