Tisk…

Zinc chloride hydrate ≥99.999% (metals basis), Puratronic®

Dodavatel: Thermo Fisher Scientific
Nebezpečí

35782.09 35782.18 35782.30
35782.09EA 2300 CZK
35782.09 35782.18 35782.30
Zinc chloride hydrate ≥99.999% (metals basis), Puratronic®
Zinc chloride hydrate

Purity: 99.999% CAS No.: 7646-85-7 Molecular Formula: Cl2Zn.xH2O Formula Weight: 136.29(anhy) Physical Form: -10 Mesh Crystalline Aggregates Appearance: White Odour: Odourless Merck Reference: 14,10132 UN No.: 3260 MDL No.: MFCD00011295

Vzorec: Cl₂Zn·H₂O
Storage Temperature: Ambient
MDL Number: MFCD00011295
CAS číslo: 7646-85-7
EINECS: 231-592-0
UN: 3260
ADR: 8 III
Merck Index: 14,10132

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR